gRNA Design Details

Target DNA Sequence
AGTGTACGAAGCTGCCGCTG
PAM Sequence
TGG
Target locus
Chr.13: 28061991 - 28062013 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 87.64/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FLT3
Entrez Gene ID
2322
Gene Name
fms related tyrosine kinase 3
Gene Aliases
CD135, FLK-2, FLK2, STK1
Chromosome Location
Chr.13: 28003274 - 28100592 on GRCh38
UniGene ID
Hs.507590
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_004119.2 NP_004110.2 3
NR_130706.1 3
XM_011535015.2 XP_011533317.1 3
XM_011535017.2 XP_011533319.1 2
XM_017020486.1 XP_016875975.1 3
XM_017020487.1 XP_016875976.1 3
XM_017020488.1 XP_016875977.1 3
XM_017020489.1 XP_016875978.1 3