gRNA Design Details
- Target DNA Sequence
- GGTGGACACAACTCTCCCAA
- PAM Sequence
- AGG
- Target locus
-
Chr.8: 23212118 - 23212140 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.93/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TNFRSF10A
- Entrez Gene ID
-
8797
- Gene Name
- TNF receptor superfamily member 10a
- Gene Aliases
-
APO2, CD261, DR4, TRAILR-1, TRAILR1
- Chromosome Location
-
Chr.8: 23191457 - 23225167 on GRCh38
- UniGene ID
- Hs.213467
- Species
-
Human