gRNA Design Details
- Target DNA Sequence
- TCAACACGGTGTCCAAGATC
- PAM Sequence
- CGG
- Target locus
-
Chr.19: 4365579 - 4365601 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
85.93/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SH3GL1
- Entrez Gene ID
-
6455
- Gene Name
- SH3 domain containing GRB2 like 1, endophilin A2
- Gene Aliases
-
CNSA1, EEN, SH3D2B, SH3P8
- Chromosome Location
-
Chr.19: 4360367 - 4400568 on GRCh38
- UniGene ID
- Hs.490415
- Species
-
Human