gRNA Design Details
- Target DNA Sequence
- ACGTTTGTGGAGACGGAATC
- PAM Sequence
- TGG
- Target locus
-
Chr.13: 94612227 - 94612249 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
94.27/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GPR180
- Entrez Gene ID
-
160897
- Gene Name
- G protein-coupled receptor 180
- Gene Aliases
-
ITR
- Chromosome Location
-
Chr.13: 94601850 - 94634645 on GRCh38
- UniGene ID
- Hs.439363
- Species
-
Human