gRNA Design Details
- Target DNA Sequence
- TCACAGGAGGTGTTGGAGGT
- PAM Sequence
- GGG
- Target locus
-
Chr.17: 35997717 - 35997739 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
83.93/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CCL15
- Entrez Gene ID
-
6359
- Gene Name
- C-C motif chemokine ligand 15
- Gene Aliases
-
HCC-2, HMRP-2B, LKN-1, LKN1, MIP-1 delta, MIP-1D, MIP-5, MRP-2B, NCC-3, NCC3, SCYA15, SCYL3, SY15
- Chromosome Location
-
Chr.17: 35997582 - 36002038 on GRCh38
- UniGene ID
- Hs.272493
- Species
-
Human