gRNA Design Details

Target DNA Sequence
AGCTTTGTATACGCTGCCAT
PAM Sequence
AGG
Target locus
Chr.20: 44978445 - 44978467 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 90.71/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
STK4
Entrez Gene ID
6789
Gene Name
serine/threonine kinase 4
Gene Aliases
KRS2, MST1, YSK3
Chromosome Location
Chr.20: 44966474 - 45079977 on GRCh38
UniGene ID
Hs.472838
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_006282.3 NP_006273.1 3
XM_005260530.2 XP_005260587.1 3
XM_005260531.3 XP_005260588.1 3
XM_005260532.3 XP_005260589.1 4
XM_005260533.2 XP_005260590.1 3
XM_011529018.2 XP_011527320.1 4
XM_011529020.2 XP_011527322.1 3
XM_017028029.1 XP_016883518.1 4
XM_017028030.1 XP_016883519.1 5
XM_017028031.1 XP_016883520.1 5
XM_017028032.1 XP_016883521.1 4
XM_017028033.1 XP_016883522.1 3