Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_006282.3 | NP_006273.1 | 4 |
XM_005260530.2 | XP_005260587.1 | 4 | |
XM_005260531.3 | XP_005260588.1 | 4 | |
XM_005260532.3 | XP_005260589.1 | 5 | |
XM_005260533.2 | XP_005260590.1 | 4 | |
XM_011529018.2 | XP_011527320.1 | 5 | |
XM_011529020.2 | XP_011527322.1 | 4 | |
XM_017028029.1 | XP_016883518.1 | 5 | |
XM_017028030.1 | XP_016883519.1 | 6 | |
XM_017028031.1 | XP_016883520.1 | 6 | |
XM_017028032.1 | XP_016883521.1 | 5 | |
XM_017028033.1 | XP_016883522.1 | 4 |