gRNA Design Details
- Target DNA Sequence
- AACTGTTCATCACTATGTCC
- PAM Sequence
- CGG
- Target locus
-
Chr.12: 4445532 - 4445554 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.16/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF6
- Entrez Gene ID
-
2251
- Gene Name
- fibroblast growth factor 6
- Gene Aliases
-
HBGF-6, HST2
- Chromosome Location
-
Chr.12: 4434142 - 4445799 on GRCh38
- UniGene ID
- Hs.166015
- Species
-
Human