gRNA Design Details
- Target DNA Sequence
- GAAATTTCCACTGTGGAGCG
- PAM Sequence
- AGG
- Target locus
-
Chr.12: 4444206 - 4444228 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.66/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF6
- Entrez Gene ID
-
2251
- Gene Name
- fibroblast growth factor 6
- Gene Aliases
-
HBGF-6, HST2
- Chromosome Location
-
Chr.12: 4434142 - 4445799 on GRCh38
- UniGene ID
- Hs.166015
- Species
-
Human