gRNA Design Details
- Target DNA Sequence
- TGCCTTAAGCCAATGATGCG
- PAM Sequence
- GGG
- Target locus
-
Chr.5: 58494726 - 58494748 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
91.45/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GAPT
- Entrez Gene ID
-
202309
- Gene Name
- GRB2-binding adaptor protein, transmembrane
- Gene Aliases
-
C5orf29
- Chromosome Location
-
Chr.5: 58491435 - 58496358 on GRCh38
- UniGene ID
- Hs.547697
- Species
-
Human