gRNA Design Details
- Target DNA Sequence
- GCATGTCCAGTCGGATCGTT
- PAM Sequence
- GGG
- Target locus
-
Chr.16: 2785924 - 2785946 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
96.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS33
- Entrez Gene ID
-
260429
- Gene Name
- protease, serine 33
- Gene Aliases
-
EOS
- Chromosome Location
-
Chr.16: 2783953 - 2787759 on GRCh38
- UniGene ID
- Hs.280658
- Species
-
Human