gRNA Design Details

Target DNA Sequence
CGAACCCCTCTGAGAATCGT
PAM Sequence
GGG
Target locus
Chr.3: 46833646 - 46833668 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 89.21/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PRSS42
Entrez Gene ID
339906
Gene Name
protease, serine 42
Gene Aliases
TESSP2
Chromosome Location
Chr.3: 46830404 - 46834095 on GRCh38
UniGene ID
Hs.585127
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_182702.1 NP_874361.1 2