gRNA Design Details
- Target DNA Sequence
- CGAACCCCTCTGAGAATCGT
- PAM Sequence
- GGG
- Target locus
-
Chr.3: 46833646 - 46833668 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
89.21/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS42
- Entrez Gene ID
-
339906
- Gene Name
- protease, serine 42
- Gene Aliases
-
TESSP2
- Chromosome Location
-
Chr.3: 46830404 - 46834095 on GRCh38
- UniGene ID
- Hs.585127
- Species
-
Human