gRNA Design Details
- Target DNA Sequence
- CTCCCATCTTGACACTGTAA
- PAM Sequence
- TGG
- Target locus
-
Chr.3: 46833178 - 46833200 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.13/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS42
- Entrez Gene ID
-
339906
- Gene Name
- protease, serine 42
- Gene Aliases
-
TESSP2
- Chromosome Location
-
Chr.3: 46830404 - 46834095 on GRCh38
- UniGene ID
- Hs.585127
- Species
-
Human