gRNA Design Details

Target DNA Sequence
TTCTGCACGGGCACACTTAA
PAM Sequence
AGG
Target locus
Chr.12: 69571285 - 69571307 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 95.91/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FRS2
Entrez Gene ID
10818
Gene Name
fibroblast growth factor receptor substrate 2
Gene Aliases
FRS1A, FRS2A, FRS2alpha, SNT, SNT-1, SNT1
Chromosome Location
Chr.12: 69470349 - 69579793 on GRCh38
UniGene ID
Hs.593446
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001042555.2 NP_001036020.1 8
NM_001278351.1 NP_001265280.1 9
NM_001278353.1 NP_001265282.1 9
NM_001278354.1 NP_001265283.1 8
NM_001278355.1 NP_001265284.1 8
NM_001278356.1 NP_001265285.1 7
NM_001278357.1 NP_001265286.1 5
NM_006654.4 NP_006645.3 8
XM_017018717.1 XP_016874206.1 9
XM_017018718.1 XP_016874207.1 9
XM_017018719.1 XP_016874208.1 4