gRNA Design Details
- Target DNA Sequence
- CAAGGTTGAGTGCACTCAAT
- PAM Sequence
- TGG
- Target locus
-
Chr.4: 15936368 - 15936390 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
90.33/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGFBP1
- Entrez Gene ID
-
9982
- Gene Name
- fibroblast growth factor binding protein 1
- Gene Aliases
-
FGF-BP, FGF-BP1, FGFBP, FGFBP-1, HBP17
- Chromosome Location
-
Chr.4: 15935569 - 15938740 on GRCh38
- UniGene ID
- Hs.1690
- Species
-
Human