gRNA Design Details

Target DNA Sequence
AGTGGTCGAAGGTGTCAAAC
PAM Sequence
TGG
Target locus
Chr.12: 69570490 - 69570512 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 88.93/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FRS2
Entrez Gene ID
10818
Gene Name
fibroblast growth factor receptor substrate 2
Gene Aliases
FRS1A, FRS2A, FRS2alpha, SNT, SNT-1, SNT1
Chromosome Location
Chr.12: 69470349 - 69579793 on GRCh38
UniGene ID
Hs.593446
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001042555.2 NP_001036020.1 7
NM_001278351.1 NP_001265280.1 8
NM_001278353.1 NP_001265282.1 8
NM_001278354.1 NP_001265283.1 7
NM_001278355.1 NP_001265284.1 7
NM_001278356.1 NP_001265285.1 6
NM_001278357.1 NP_001265286.1 4
NM_006654.4 NP_006645.3 7
XM_017018717.1 XP_016874206.1 8
XM_017018718.1 XP_016874207.1 8
XM_017018719.1 XP_016874208.1 3