gRNA Design Details
- Target DNA Sequence
- ACACAGTGCAGATGAATTCC
- PAM Sequence
- AGG
- Target locus
-
Chr.20: 35437853 - 35437875 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.01/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GDF5
- Entrez Gene ID
-
8200
- Gene Name
- growth differentiation factor 5
- Gene Aliases
-
BDA1C, BMP-14, BMP14, CDMP1, LAP-4, LAP4, OS5, SYM1B, SYNS2
- Chromosome Location
-
Chr.20: 35433347 - 35454746 on GRCh38
- UniGene ID
- Hs.1573
- Species
-
Human