gRNA Design Details
- Target DNA Sequence
- AATGCCAATGCCAGGGCAAA
- PAM Sequence
- GGG
- Target locus
-
Chr.20: 35437696 - 35437718 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
85.36/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GDF5
- Entrez Gene ID
-
8200
- Gene Name
- growth differentiation factor 5
- Gene Aliases
-
BDA1C, BMP-14, BMP14, CDMP1, LAP-4, LAP4, OS5, SYM1B, SYNS2
- Chromosome Location
-
Chr.20: 35433347 - 35454746 on GRCh38
- UniGene ID
- Hs.1573
- Species
-
Human