gRNA Design Details

Target DNA Sequence
GGCCAGAAGGTTTTCAAAGT
PAM Sequence
GGG
Target locus
Chr.9: 129022355 - 129022377 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 86.26/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SH3GLB2
Entrez Gene ID
56904
Gene Name
SH3 domain containing GRB2 like endophilin B2
Gene Aliases
PP6569, PP9455, RRIG1
Chromosome Location
Chr.9: 129007792 - 129028353 on GRCh38
UniGene ID
Hs.460238
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001287045.1 NP_001273974.1 2
NM_001287046.1 NP_001273975.1 2
NM_020145.3 NP_064530.1 2
XM_005252098.2 XP_005252155.1 2
XM_005252100.2 XP_005252157.1 2
XM_006717188.2 XP_006717251.1 2
XM_006717189.2 XP_006717252.1 2
XM_006717190.1 XP_006717253.1 2
XM_006717191.1 XP_006717254.1 2