gRNA Design Details
- Target DNA Sequence
- CGAAGGTGTCTGTCTCCACT
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 171449154 - 171449176 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF18
- Entrez Gene ID
-
8817
- Gene Name
- fibroblast growth factor 18
- Gene Aliases
-
FGF-18, ZFGF5
- Chromosome Location
-
Chr.5: 171419663 - 171457626 on GRCh38
- UniGene ID
- Hs.87191
- Species
-
Human