gRNA Design Details
- Target DNA Sequence
- CAACGTCTATGACCTCCTAA
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 63917792 - 63917814 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
90.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GH1
- Entrez Gene ID
-
2688
- Gene Name
- growth hormone 1
- Gene Aliases
-
GH, GH-N, GHB5, GHN, IGHD1B, hGH-N
- Chromosome Location
-
Chr.17: 63917193 - 63918852 on GRCh38
- UniGene ID
- Hs.655229
- Species
-
Human