gRNA Design Details

Target DNA Sequence
GGATGGGCTAGTTAAGTCGT
PAM Sequence
TGG
Target locus
Chr.8: 10531324 - 10531346 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 95.64/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PRSS55
Entrez Gene ID
203074
Gene Name
protease, serine 55
Gene Aliases
CT153, T-SP1, TSP1, UNQ9391
Chromosome Location
Chr.8: 10525331 - 10554166 on GRCh38
UniGene ID
Hs.591394
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001197020.1 NP_001183949.1 3
NM_198464.3 NP_940866.2 3
XM_005272372.3 XP_005272429.2 3
XM_011543814.2 XP_011542116.2 3
XM_017013182.1 XP_016868671.1 3