gRNA Design Details
- Target DNA Sequence
- ACTATTTATGACTACGTGCG
- PAM Sequence
- GGG
- Target locus
-
Chr.14: 74684542 - 74684564 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
73.25/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- AREL1
- Entrez Gene ID
-
9870
- Gene Name
- apoptosis resistant E3 ubiquitin protein ligase 1
- Gene Aliases
-
FIEL1, KIAA0317
- Chromosome Location
-
Chr.14: 74661252 - 74713118 on GRCh38
- UniGene ID
- Hs.497417
- Species
-
Human