gRNA Design Details
- Target DNA Sequence
- AATGAAGACCGTGAGCGCCG
- PAM Sequence
- AGG
- Target locus
-
Chr.12: 84941849 - 84941871 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
91.88/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Arel1
- Entrez Gene ID
-
68497
- Gene Name
- apoptosis resistant E3 ubiquitin protein ligase 1
- Gene Aliases
-
1110018G07Rik, AI649076, AW610792, Kiaa0317, mKIAA0317
- Chromosome Location
-
Chr.12: 84918140 - 84970886 on GRCm38
- UniGene ID
- Mm.24446
- Species
-
Mouse