gRNA Design Details
- Target DNA Sequence
- ACTCATTGATGATGATATCG
- PAM Sequence
- GGG
- Target locus
-
Chr.11: 113931844 - 113931866 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.61/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HTR3B
- Entrez Gene ID
-
9177
- Gene Name
- 5-hydroxytryptamine receptor 3B
- Gene Aliases
-
5-HT3B
- Chromosome Location
-
Chr.11: 113904796 - 113949682 on GRCh38
- UniGene ID
- Hs.241377
- Species
-
Human