gRNA Design Details
- Target DNA Sequence
- CAAGTGTAATGGCTCCCCTG
- PAM Sequence
- TGG
- Target locus
-
Chr.11: 113904941 - 113904963 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.11/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HTR3B
- Entrez Gene ID
-
9177
- Gene Name
- 5-hydroxytryptamine receptor 3B
- Gene Aliases
-
5-HT3B
- Chromosome Location
-
Chr.11: 113904796 - 113949682 on GRCh38
- UniGene ID
- Hs.241377
- Species
-
Human