gRNA Design Details
- Target DNA Sequence
- GCATAAACCACCGTTGTGGC
- PAM Sequence
- AGG
- Target locus
-
Chr.15: 88909883 - 88909905 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
95.36/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MFGE8
- Entrez Gene ID
-
4240
- Gene Name
- milk fat globule-EGF factor 8 protein
- Gene Aliases
-
BA46, EDIL1, HMFG, HsT19888, MFG-E8, MFGM, OAcGD3S, SED1, SPAG10, hP47
- Chromosome Location
-
Chr.15: 88898683 - 88913469 on GRCh38
- UniGene ID
- Hs.3745
- Species
-
Human