gRNA Design Details
- Target DNA Sequence
- GTCATCGTAGCTGCAGACAC
- PAM Sequence
- AGG
- Target locus
-
Chr.16: 1792265 - 1792287 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
90.28/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IGFALS
- Entrez Gene ID
-
3483
- Gene Name
- insulin like growth factor binding protein acid labile subunit
- Gene Aliases
-
ACLSD, ALS
- Chromosome Location
-
Chr.16: 1790413 - 1794908 on GRCh38
- UniGene ID
- Hs.839
- Species
-
Human