gRNA Design Details

Target DNA Sequence
TGCTGGTATCAGGCTCACTC
PAM Sequence
GGG
Target locus
Chr.16: 1792446 - 1792468 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 85.38/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
IGFALS
Entrez Gene ID
3483
Gene Name
insulin like growth factor binding protein acid labile subunit
Gene Aliases
ACLSD, ALS
Chromosome Location
Chr.16: 1790413 - 1794908 on GRCh38
UniGene ID
Hs.839
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001146006.1 NP_001139478.1 2