gRNA Design Details
- Target DNA Sequence
- GTCGAACACAGTGGTACCTG
- PAM Sequence
- AGG
- Target locus
-
Chr.15: 49424509 - 49424531 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.75/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF7
- Entrez Gene ID
-
2252
- Gene Name
- fibroblast growth factor 7
- Gene Aliases
-
HBGF-7, KGF
- Chromosome Location
-
Chr.15: 49423178 - 49487326 on GRCh38
- UniGene ID
- Hs.567268
- Species
-
Human