gRNA Design Details

Target DNA Sequence
ATCTGTAGAGCAAAGTTGGC
PAM Sequence
AGG
Target locus
Chr.15: 49424324 - 49424346 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 86.42/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FGF7
Entrez Gene ID
2252
Gene Name
fibroblast growth factor 7
Gene Aliases
HBGF-7, KGF
Chromosome Location
Chr.15: 49423178 - 49487326 on GRCh38
UniGene ID
Hs.567268
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_002009.3 NP_002000.1 2