gRNA Design Details
- Target DNA Sequence
- CTTGCGCGGCTTTGCTGTAC
- PAM Sequence
- TGG
- Target locus
-
Chr.6: 35214481 - 35214503 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SCUBE3
- Entrez Gene ID
-
222663
- Gene Name
- signal peptide, CUB domain and EGF like domain containing 3
- Gene Aliases
-
CEGF3
- Chromosome Location
-
Chr.6: 35214047 - 35253079 on GRCh38
- UniGene ID
- Hs.12923
- Species
-
Human