gRNA Design Details

Target DNA Sequence
ATCCATCATAGCAGGTACAC
PAM Sequence
CGG
Target locus
Chr.6: 35228684 - 35228706 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 89.6/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SCUBE3
Entrez Gene ID
222663
Gene Name
signal peptide, CUB domain and EGF like domain containing 3
Gene Aliases
CEGF3
Chromosome Location
Chr.6: 35214047 - 35253079 on GRCh38
UniGene ID
Hs.12923
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001303136.1 NP_001290065.1 3
NM_152753.3 NP_689966.2 3
XM_005248943.1 XP_005249000.1 3
XM_005248947.1 XP_005249004.1 3
XM_011514404.2 XP_011512706.1 3