gRNA Design Details

Target DNA Sequence
CTCATACACTTTCCGTGCAT
PAM Sequence
GGG
Target locus
Chr.20: 20045392 - 20045414 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 89.67/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CRNKL1
Entrez Gene ID
51340
Gene Name
crooked neck pre-mRNA splicing factor 1
Gene Aliases
CLF, CRN, Clf1, HCRN, MSTP021, SYF3
Chromosome Location
Chr.20: 20034361 - 20056046 on GRCh38
UniGene ID
Hs.171342
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001278625.1 NP_001265554.1 7
NM_001278626.1 NP_001265555.1 7
NM_001278627.1 NP_001265556.1 7
NM_001278628.1 NP_001265557.1 6
NM_016652.5 NP_057736.4 7