gRNA Design Details

Target DNA Sequence
GACTCTGATCTTCGGCAAGG
PAM Sequence
TGG
Target locus
Chr.17: 57105762 - 57105784 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 92.22/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
AKAP1
Entrez Gene ID
8165
Gene Name
A-kinase anchoring protein 1
Gene Aliases
AKAP, AKAP121, AKAP149, AKAP84, D-AKAP1, PPP1R43, PRKA1, SAKAP84, TDRD17
Chromosome Location
Chr.17: 57085192 - 57121349 on GRCh38
UniGene ID
Hs.463506
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001242902.1 NP_001229831.1 3
NM_001242903.1 NP_001229832.1 3
NM_003488.3 NP_003479.1 2
XM_005257707.2 XP_005257764.1 3
XM_005257709.2 XP_005257766.1 2
XM_017025189.1 XP_016880678.1 3
XM_017025190.1 XP_016880679.1 2
XM_017025191.1 XP_016880680.1 2