gRNA Design Details
- Target DNA Sequence
- TGTCGTTCCTTGAAGAGGCG
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 3611245 - 3611267 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.67/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MEGF6
- Entrez Gene ID
-
1953
- Gene Name
- multiple EGF like domains 6
- Gene Aliases
-
EGFL3
- Chromosome Location
-
Chr.1: 3487942 - 3624757 on GRCh38
- UniGene ID
- Hs.593645
- Species
-
Human