gRNA Design Details

Target DNA Sequence
CCGTCTACTACATGGGCTAC
PAM Sequence
AGG
Target locus
Chr.1: 3595423 - 3595445 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 89.53/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
MEGF6
Entrez Gene ID
1953
Gene Name
multiple EGF like domains 6
Gene Aliases
EGFL3
Chromosome Location
Chr.1: 3487942 - 3624757 on GRCh38
UniGene ID
Hs.593645
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001409.3 NP_001400.3 3
XM_011540885.1 XP_011539187.1 4
XM_011540886.1 XP_011539188.1 4
XM_011540888.2 XP_011539190.1 4
XM_017000533.1 XP_016856022.1 4