gRNA Design Details
- Target DNA Sequence
- TGGTCGCTGTCCACATGCAC
- PAM Sequence
- AGG
- Target locus
-
Chr.10: 47300872 - 47300894 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
85.23/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GDF10
- Entrez Gene ID
-
2662
- Gene Name
- growth differentiation factor 10
- Gene Aliases
-
BMP-3b, BMP3B
- Chromosome Location
-
Chr.10: 47300224 - 47313577 on GRCh38
- UniGene ID
- Hs.2171
- Species
-
Human