gRNA Design Details
- Target DNA Sequence
- GTGGTAGACGAAGCAGTCGT
- PAM Sequence
- TGG
- Target locus
-
Chr.2: 105308158 - 105308180 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.1/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TGFBRAP1
- Entrez Gene ID
-
9392
- Gene Name
- transforming growth factor beta receptor associated protein 1
- Gene Aliases
-
TRAP-1, TRAP1, VPS3
- Chromosome Location
-
Chr.2: 105249404 - 105329714 on GRCh38
- UniGene ID
- Hs.446350
- Species
-
Human