gRNA Design Details
- Target DNA Sequence
- GCACTTCAAGGTGCTGCGTG
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 19024343 - 19024365 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
82.48/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GRAP
- Entrez Gene ID
-
10750
- Gene Name
- GRB2-related adaptor protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 19020656 - 19051352 on GRCh38
- UniGene ID
- Hs.567416
- Species
-
Human