gRNA Design Details

Target DNA Sequence
CTCATACACGCTCGTGTCCA
PAM Sequence
TGG
Target locus
Chr.2: 97734556 - 97734578 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 94/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ZAP70
Entrez Gene ID
7535
Gene Name
zeta chain of T cell receptor associated protein kinase 70
Gene Aliases
ADMIO2, IMD48, SRK, STCD, STD, TZK, ZAP-70
Chromosome Location
Chr.2: 97712030 - 97744327 on GRCh38
UniGene ID
Hs.234569
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001079.3 NP_001070.2 9
NM_207519.1 NP_997402.1 1
XM_017004867.1 XP_016860356.1 11
XM_017004868.1 XP_016860357.1 11
XM_017004869.1 XP_016860358.1 11
XM_017004870.1 XP_016860359.1 11