gRNA Design Details
- Target DNA Sequence
- ACGGTCTGGTTGGCTCCACA
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 49502162 - 49502184 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.99/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- NGFR
- Entrez Gene ID
-
4804
- Gene Name
- nerve growth factor receptor
- Gene Aliases
-
CD271, Gp80-LNGFR, TNFRSF16, p75(NTR), p75NTR
- Chromosome Location
-
Chr.17: 49495293 - 49515020 on GRCh38
- UniGene ID
- Hs.415768
- Species
-
Human