gRNA Design Details

Target DNA Sequence
CTCGTAAGGGAAGAGGTCTT
PAM Sequence
TGG
Target locus
Chr.1: 156745110 - 156745132 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 87.48/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
HDGF
Entrez Gene ID
3068
Gene Name
hepatoma-derived growth factor
Gene Aliases
HMG1L2
Chromosome Location
Chr.1: 156742107 - 156752448 on GRCh38
UniGene ID
Hs.743948
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001126050.1 NP_001119522.1 3
NM_001126051.1 NP_001119523.1 3
NM_001319186.1 NP_001306115.1 3
NM_001319187.1 NP_001306116.1 3
NM_001319188.1 NP_001306117.1 4
NM_004494.2 NP_004485.1 3
NR_135008.1 3
XM_011509453.2 XP_011507755.2 3
XM_011509454.2 XP_011507756.1 3
XM_011509455.2 XP_011507757.1 3