gRNA Design Details
- Target DNA Sequence
- GAAGAATGTACGCTTTGACC
- PAM Sequence
- AGG
- Target locus
-
Chr.12: 51074023 - 51074045 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.33/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CSRNP2
- Entrez Gene ID
-
81566
- Gene Name
- cysteine and serine rich nuclear protein 2
- Gene Aliases
-
C12orf2, C12orf22, FAM130A1, PPP1R72, TAIP-12
- Chromosome Location
-
Chr.12: 51061205 - 51083671 on GRCh38
- UniGene ID
- Hs.524425
- Species
-
Human