gRNA Design Details
- Target DNA Sequence
- TCCCAAGTACCAAGATACAA
- PAM Sequence
- CGG
- Target locus
-
Chr.4: 151282247 - 151282269 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
92.42/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS48
- Entrez Gene ID
-
345062
- Gene Name
- protease, serine 48
- Gene Aliases
-
ESSPL
- Chromosome Location
-
Chr.4: 151276482 - 151291856 on GRCh38
- UniGene ID
- Hs.651266
- Species
-
Human