gRNA Design Details

Target DNA Sequence
GATCTTCCATGTCCAGTCTC
PAM Sequence
TGG
Target locus
Chr.12: 51976472 - 51976494 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 86.15/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ACVR1B
Entrez Gene ID
91
Gene Name
activin A receptor type 1B
Gene Aliases
ACTRIB, ACVRLK4, ALK4, SKR2
Chromosome Location
Chr.12: 51951667 - 51997079 on GRCh38
UniGene ID
Hs.438918
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_004302.4 NP_004293.1 3
NM_020327.3 NP_064732.3 3
NM_020328.3 NP_064733.3 3
XM_011538966.2 XP_011537268.1 3
XM_011538967.2 XP_011537269.1 3
XM_017020199.1 XP_016875688.1 3
XM_017020200.1 XP_016875689.1 4
XM_017020201.1 XP_016875690.1 3