gRNA Design Details

Target DNA Sequence
TCTGTTGCAGGTGATAACGC
PAM Sequence
AGG
Target locus
Chr.5: 147323958 - 147323980 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 89.85/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
STK32A
Entrez Gene ID
202374
Gene Name
serine/threonine kinase 32A
Gene Aliases
YANK1
Chromosome Location
Chr.5: 147234963 - 147401996 on GRCh38
UniGene ID
Hs.585069
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001112724.1 NP_001106195.1 5
NM_001287740.1 NP_001274669.1 5
NM_145001.3 NP_659438.1 5
XM_011537576.1 XP_011535878.1 4
XM_011537577.2 XP_011535879.1 5
XM_011537579.2 XP_011535881.1 5
XM_017009213.1 XP_016864702.1 5