gRNA Design Details
- Target DNA Sequence
- GCGACGGCTATGATTACGAT
- PAM Sequence
- GGG
- Target locus
-
Chr.17: 58006479 - 58006501 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
97/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SRSF1
- Entrez Gene ID
-
6426
- Gene Name
- serine and arginine rich splicing factor 1
- Gene Aliases
-
ASF, SF2, SF2p33, SFRS1, SRp30a
- Chromosome Location
-
Chr.17: 58000919 - 58007346 on GRCh38
- UniGene ID
- Hs.68714
- Species
-
Human