gRNA Design Details
- Target DNA Sequence
- GATCACTTTGACAGGGAGGT
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 48731268 - 48731290 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
88.42/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- LHCGR
- Entrez Gene ID
-
3973
- Gene Name
- luteinizing hormone/choriogonadotropin receptor
- Gene Aliases
-
HHG, LCGR, LGR2, LH/CG-R, LH/CGR, LHR, LHRHR, LSH-R, ULG5
- Chromosome Location
-
Chr.2: 48686774 - 48755741 on GRCh38
- UniGene ID
- Hs.468490
- Species
-
Human